Learn More
ACROBiosystems Human CD39 / ENTPD1 Protein, His Tag (active enzyme)
Description
CD39 is also known as Ectonucleoside triphosphate diphosphohydrolase 1, ENTPD1, NTPDase 1, Ecto-ATPDase 1, in the nervous system, could hydrolyze ATP and other nucleotides to regulate purinergic neurotransmission. Could also be implicated in the prevention of platelet aggregation by hydrolyzing platelet-activating ADP to AMP. Hydrolyzes ATP and ADP equally well. NTPDase-1 was originally described as CD39, a B lymphocyte cell surface marker, but it is also present on the surface of natural killer cells, T cells, and some endothelial cells.Regulatory T cells(Tregs) mediate immunosuppression through multiple, non-redundant, cell-contact dependent and independent mechanisms, a growing body of evidence suggests an important role for the CD39-CD73-adenosine pathway. CD39 ectonucleotidase is the rate-limiting enzyme of a cascade leading to the generation of suppressive adenosine that alters CD4 and CD8 T cell and natural killer cell antitumor activities.
Spécification
Spécification
| Numéro d’adhésion | NP_001767.3 |
| Poids moléculaire | 52.3 kDa |
| Quantité | 1 mg |
| Source | Human CD39, His Tag (CD9-H52H4) is expressed from human 293 cells (HEK293). It contains AA Thr 38 - Val 478 (Accession # P49961-1). |
| Immunogène | Thr 38 - Val 478 |
| Conditions de stockage | -20°C |
| Alias de gène | CD39,ENTPD1,NTPDase 1,Entpd1,Ecto-ATPDase 1,Ecto-ATPase 1 |
| Nom usuel | CD39 |
| Conjugué | Unconjugated |
| Espèces | Human |
| Afficher plus |
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.