Learn More
R&D Systems™ Human CLEC3A (O75596) VersaClone cDNA
Description
This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.
Informations sur la commande
Ships at ambient temperature
Spécification
Spécification
| Expression | CLEC3A |
| Numéro d’adhésion | O75596 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formule | cDNA is provided in 10mM Tris-Cl, pH 8.347 |
| Type d’insert | 607bp |
| Identification génétique (Entrez) | 10143 |
| Alias de gène | Cartilage-derived C-type lectin, CLECSF1, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 1 (cartilage-derived), C-type lectin domain family 3 member A, C-type lectin domain family 3, member A, C-type lectin superfamily member 1, C-type lectin, superfamily member 1, MGC129558, MGC129559 |
| Contenu et stockage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Espèces | Human |
| Quantité | 10 μg |
For Reasearch Use Only. Not for use in Humans.