Learn More
R&D Systems™ Human IgG B Cell ELISpot Development Module, 5 Plate
Code produit 18471540
Change view
Click to view available options
Quantité:
1 kit
Conditionnement:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18471540 1 kit 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 18471540 Fournisseur R&D Systems™ Code fournisseur SELB002
Sufficient for at least five 96-well plates
TRUSTED_SUSTAINABILITY
Spécification
Quantité 1 kit
Contenu et stockage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.