missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NAGA (aa 30-161) Control Fragment Recombinant Protein

Code produit 30196276
Change view
Click to view available options
Quantité:
100 μl
Conditionnement:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30196276 100 μl 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 30196276 Fournisseur Invitrogen™ Code fournisseur RP91478

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (88%), Rat (88%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-51433 (PA5-51433. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

NAGA (N-acetylgalactosaminidase, alpha), also known as alpha-galactosidase B or GALB, is a 411 lysosomal protein belonging to the glycosyl hydrolase 27 family that may exist as a homodimer and plays a critical role in glycolipid breakdown. NAGA encodes alpha-N-acetylgalactosaminidase, a lysosomal enzyme, which cleaves alpha-N-acetylgalactosaminyl groups from glycoconjugates. Mapping to human chromosome 22q13. 2, NAGA defects are the cause of an autosomal recessive disorder with three phenotypes, known as Schindler disease (types I, II and III) or NAGA deficiency (types I, II and III). Characterized by neurologic manifestations that range in severity, Schindler disease type I is the most severe form, followed by type III, which may have mild-to-moderate effects. Schindler disease type II, also known as Kanzaki disease, is characterized by mild intellectual impairment and angiokeratoma corporis diffusum.
TRUSTED_SUSTAINABILITY

Spécification

Numéro d’adhésion P17050
Concentration ≥5.0 mg/mL
À utiliser avec (application) Blocking Assay, Control
Formule 1 M urea, PBS with no preservative; pH 7.4
Identification génétique (Entrez) 4668
Nom Human NAGA (aa 30-161) Control Fragment
Quantité 100 μl
État réglementaire RUO
Alias de gène Acetylgalactosaminidase, alpha-N- (alpha-galactosidase B); alpha-galactosidase B; Alpha-N-acetylgalactosaminidase; D22S674; EC 3.2.1; EC 3.2.1.49; galactosidasealphaB; galactosidase-alphaB; GALB; N-acetyl galactosaminidase, alpha; N-acetylgalactosaminidase, alpha-; Naga
Nom usuel NAGA
Symbole de gène(s) NAGA
Conjugué Unconjugated
Espèces Human
Recombinant Recombinant
Marqueur de protéine His-ABP-tag
Séquence WLAWERFRCNINCDEDPKNCISEQLFMEMADRMAQDGWRDMGYTYLNIDDCWIGGRDASGRLMPDPKRFPHGIPFLADYVHSLGLKLGIYADMGNFTCMGYPGTTLDKVVQDAQTFAEWKVDMLKLDGCFST
Contenu et stockage -20°C, Avoid Freeze/Thaw Cycles
Système d’expression E. coli
Forme Liquid
Pureté ou qualité >80% by SDS-PAGE and Coomassie blue staining
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.