Learn More
STEMCELL Technologies Human pan IFN-alpha ELISA Kit
Description
- The Human pan Interferon-alpha (IFN-α) ELISA Kit is designed for the quantitative detection and measurement of human IFN-α in biological fluids such as serum, plasma, and cell culture supernatants
- IFN-α is primarily produced by macrophages resulting from viral infections and activates anti-viral and immunomodulatory activities in target cells
- This kit is specific for subtypes 1/13, 2, 4, 5, 6, 7, 8, 10, 14, 16, and 17
- The assay is based on the sandwich ELISA method, in which samples are added to ELISA strip plates pre-coated with capture antibodies specific for the cytokine
- The captured cytokine is detected by addition of a biotinylated detection antibody, followed by streptavidin-horseradish peroxidase, which binds the biotinylated antibody
- Addition of the chromogenic enzyme substrate 3,3',5,5' tetramethylbenzidine (TMB) results in a colored product with an intensity directly proportional to the concentration of cytokine in the sample
Specifications
Specifications
| Description | Dendritic Cells; Macrophages |
| À utiliser avec (application) | ELISA |
| Quantité | 2 plaques |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?