Learn More
STEMCELL Technologies Human pan IFN-alpha ELISA Kit
Code produit 17188001
Encompass_Preferred
Change view
Click to view available options
Quantité:
10 plaques
2 plaques
Conditionnement:
10 pièces
2 pièces
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
17188001 2 plaques 2 pièces
17198001 10 plaques 10 pièces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 17188001 Fournisseur STEMCELL Technologies Code fournisseur 02000
For detection and measurement of pan human interferon-alpha
  • The Human pan Interferon-alpha (IFN-α) ELISA Kit is designed for the quantitative detection and measurement of human IFN-α in biological fluids such as serum, plasma, and cell culture supernatants
  • IFN-α is primarily produced by macrophages resulting from viral infections and activates anti-viral and immunomodulatory activities in target cells
  • This kit is specific for subtypes 1/13, 2, 4, 5, 6, 7, 8, 10, 14, 16, and 17
  • The assay is based on the sandwich ELISA method, in which samples are added to ELISA strip plates pre-coated with capture antibodies specific for the cytokine
  • The captured cytokine is detected by addition of a biotinylated detection antibody, followed by streptavidin-horseradish peroxidase, which binds the biotinylated antibody
  • Addition of the chromogenic enzyme substrate 3,3',5,5' tetramethylbenzidine (TMB) results in a colored product with an intensity directly proportional to the concentration of cytokine in the sample
Specifications
Description Dendritic Cells; Macrophages
À utiliser avec (application) ELISA
Quantité 2 plaques