Learn More
R&D Systems™ Human PANK2 (NP_705902.2) VersaClone cDNA
Spécification
Spécification
| Expression | PANK2 |
| Numéro d’adhésion | NP_705902.2 |
| Identification génétique (Entrez) | 80025 |
| Alias de gène | C20orf48, EC 2.7.1.33, FLJ11729, FLJ17232, Hallervorden-Spatz syndrome, HARPFLJ11729, hPanK2, HSS, MGC15053, NBIA1, pantothenate kinase 2, pantothenate kinase 2, mitochondrial, Pantothenic acid kinase 2, PKAN, PKANneurodegeneration with brain iron accumulation 1 (Hallervorden-Spatz syndrome) |
| Contenu et stockage | Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Espèces | Human |
| Quantité | 10 μg |