Learn More
STEMCELL Technologies Human Recombinant EPO
Description
- Erythropoietin (EPO) is a glycoprotein growth factor that is produced primarily in the kidney in response to hypoxia or anemia
- It is the principal physiological regulator of erythropoiesis
- EPO promotes erythropoiesis by binding to a homodimeric cell surface receptor that activates JAK2/STAT5, PI3K/AKT, and MAPK pathways, and stimulates the proliferation and differentiation of erythroid progenitor cells (Jelkmann; Kuhrt & Wojchowski)
Spécification
Spécification
| Nom | Epoetin, Erythropoietin, EP |
| Quantité | 50 μg |
| Type de cellule | Hematopoietic Stem and Progenitor Cells; Mesoderm, PSC-Derived; Pluripotent Stem Cells |
| Pureté ou qualité | ≥95% |