missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RET (NP_000314.1, 658 a.a. - 1114 a.a.) M918T mutant Partial Recombinant Protein with GST-tag expressed in baculovirus infected Sf21 cells

Code produit 16058336
Modifier l'affichage
Click to view available options
Quantité:
5 μg
Conditionnement:
5µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
16058336 5 μg 5µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 16058336 Fournisseur Abnova™ Code fournisseur P5631.5ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Voir les produits de remplacement

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Used for Func, SDS-PAGE

This gene, a member of the cadherin superfamily, encodes one of the receptor tyrosine kinases, which are cell-surface molecules that transduce signals for cell growth and differentiation. This gene plays a crucial role in neural crest development, and it can undergo oncogenic activation in vivo and in vitro by cytogenetic rearrangement. Mutations in this gene are associated with the disorders multiple endocrine neoplasia, type IIA, multiple endocrine neoplasia, type IIB, Hirschsprung disease, and medullary thyroid carcinoma. Two transcript variants encoding different isoforms have been found for this gene. Additional transcript variants have been described but their biological validity has not been confirmed. [provided by RefSeq]

Spécification

Numéro d’adhésion NP_000314.1
À utiliser avec (application) Functional Study, SDS-PAGE
Formule Liquid
Identification génétique (Entrez) 5979
Poids moléculaire 79kDa
Nom RET (M918T) (Human) Recombinant Protein
Méthode de préparation Baculovirus infected insect cell (Sf21) expression system
Méthode de purification Glutathione sepharose chromatography
Test du contrôle qualité Loading 1 ug protein in SDS-PAGE
Quantité 5 μg
Conditions de stockage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
État réglementaire RUO
Alias de gène CDHF12/HSCR1/MEN2A/MEN2B/MTC1/PTC/RET-ELE1/RET51
Nom usuel RET
Symbole de gène(s) RET
Activité biologique The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg(or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip 3000. Substrate: CSKtide. ATP: 100 uM.
Espèces Baculovirus
Recombinant Recombinant
Marqueur de protéine GST
Système d’expression Baculovirus infected insect cell (Sf21) expression system
Forme Liquid
Pureté ou qualité 92% by SDS-PAGE/CBB staining
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.