Learn More
R&D Systems™ Human Semaphorin 5A (NP_003957) VersaClone cDNA
Description
The Human Semaphorin 5A (NP_003957) VersaClone cDNA clone contains the complete ORF for Semaphorin 5A, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Semaphorin 5A insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Spécification
Spécification
| Expression | hSEMA5A |
| Numéro d’adhésion | NP_003957 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formule | cDNA is provided in 10 mM Tris-Cl |
| Type d’insert | 3238bp |
| pH | 8.5 |
| Identification génétique (Entrez) | 9037 |
| Contenu et stockage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Espèces | Human |
| Quantité | 10 μg |