Learn More
R&D Systems™ Human VSIG1 (NP_001164024) VersaClone cDNA
Description
The Human VSIG1 (NP_001164024) VersaClone cDNA clone contains the complete ORF for VSIG1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The VSIG1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Spécification
Spécification
| Expression | VSIG1 |
| Numéro d’adhésion | NP_001164024 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formule | cDNA is provided in 10 mM Tris-Cl |
| Type d’insert | 1285bp |
| pH | 8.5 |
| Identification génétique (Entrez) | 340547 |
| Alias de gène | Cell surface A33 antigen, dJ889N15.1, Glycoprotein A34, GPA34, MGC44287,1700062D20Rik, V-set and immunoglobulin domain containing 1, V-set and immunoglobulin domain-containing protein 1 |
| Contenu et stockage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Espèces | Human |
| Afficher plus |