Description
- Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity
Spécification
Spécification
| Identification génétique (Entrez) | 4803 |
| Poids moléculaire | 13 kDa, non-disulfide bonded homodimer, non-glycosylated |
| Quantité | 10 μg |
| Espèces | Human |
| Reconstitution | Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 x PBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA). |
| Contenu et stockage | Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products. |
| Activité | Typically ≤ 3 ng/mL EC50 |
| Système d’expression | Human embryonic kidney 293 (HEK293) human cell expression system |
| Forme | Lyophilized |
| Pureté ou qualité | > 95% |