Learn More
Proteintech HumanKine™ recombinant human IL-34 protein
Code produit 17610077 Tous les produits Proteintech Produits
Change view
Click to view available options
Quantité:
10 μg
100 μg
1 000 μg
Conditionnement:
1000µg
100µg
10µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
17610077 100 μg 100µg
17600077 1 000 μg 1000µg
17620077 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Code produit 17610077 Fournisseur Proteintech Code fournisseur HZ1316100UG
HumanKine™ recombinant protein
IL-34 is a 6-α-helix, proinflammatory cytokine that serves as a ligand for colony-stimulating factor-1 receptor (CSF1R). It is expressed most abundantly by spleen cells. It primarily stimulates human peripheral blood monocyte proliferation and macrophage colony formation. It is also required for Langerhans cell and microglia development. IL-34 has been shown to play a protective role in cancer and Alzheimer’s disease, but it can promote the progression of several autoimmune, inflammatory, and metabolic diseases. As such IL-34 could be used as a biomarker to mark the progression of these diseases (PMID: 22579672, 27577879, 31940023).
Spécification
Numéro d’adhésion Q6ZMJ4
Formule 50 mM Acetate pH 4.5 + 75 mM NaCl, See Certificate of Analysis for details
Identification génétique (Entrez) 146433
Poids moléculaire 68 and 38 kDa
Quantité 100 μg
Source HEK293
Conditions de stockage Lyophilized product should be stored at -20°C to -80°C until expiry date and (at Room Temperature for 2 weeks.)
Concentration en endotoxines <1 EU/μg
Réactivité spécifique Human
Recombinant Recombinant
Reconstitution Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 xPBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
Forme Lyophilized
Pureté ou qualité >95%