Learn More
Thermo Scientific™ HyPerforma™ Smartainer™ 3.0 Bioprocess Container (BPC), Aegis™ 5-14 film
GREENER_CHOICE
Code produit 17978261 Tous les produits Thermo Scientific Produits
Change view
Click to view available options
Capacité (métrique):
200 l
500 l
Type d’insert:
À soufflet horizontal
À soufflet vertical
Conditionnement:
5 pièces
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Capacité (métrique) Type d’insert unitSize
17978261 500 l À soufflet vertical 5 pièces
17958221 200 l À soufflet horizontal 5 pièces
17948221 500 l À soufflet horizontal 5 pièces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Code produit 17978261 Fournisseur Thermo Scientific™ Code fournisseur SH31188.03
Please to purchase this item. Need a web account? Register with us today!
The Thermo Scientific HyPerforma Smartainer 3.0 BPC provides end-users producing biologics or vaccine therapeutics with a large-volume, sterile liquid storage system for volumes ranging from 200 L to 3000 L—designed for the collection and storage of sterile process liquids such as cell culture media, process water or purification media. Options include temperature-control through integrated water jackets, BPC deploying hoists, as well as other options to optimize performance in your productions processes.
The Thermo Scientific HyPerforma Smartainer 3.0 BPC provides end-users producing biologics or vaccine therapeutics with a large-volume, sterile liquid storage system for volumes ranging from 200 L to 3000 L—designed for the collection and storage of sterile process liquids such as cell culture media, process water or purification media. Options include temperature-control through integrated water jackets, BPC deploying hoists, as well as other options to optimize performance in your productions processes.
TRUSTED_SUSTAINABILITY
Spécification
Capacité (métrique) 500 l
Product Type Bioprocess Container (BPC)
Type d’insert À soufflet vertical