Learn More
Thermo Scientific™ HyPerforma™ Smartainer™ 3.0 Bioprocess Container (BPC), Aegis™ 5-14 film
Code produit 31940834 Tous les produits Thermo Scientific Produits
Modifier l'affichage
Click to view available options
Capacité (métrique):
200 l
500 l
Type d’insert:
À soufflet horizontal
À soufflet vertical
Conditionnement:
1 pièce
5 pièces
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Capacité (métrique) Type d’insert Conditionnement
31940834 500 l À soufflet vertical 1 pièce
17958221 200 l À soufflet horizontal 5 pièces
17948221 500 l À soufflet horizontal 5 pièces
31940830 500 l À soufflet horizontal 1 pièce
17978261 500 l À soufflet vertical 5 pièces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Code produit 31940834 Fournisseur Thermo Scientific™ Code fournisseur SH31188.03
 Article restreint
Expédition estimée: 08-04-2027
The Thermo Scientific HyPerforma Smartainer 3.0 BPC provides end-users producing biologics or vaccine therapeutics with a large-volume, sterile liquid storage system for volumes ranging from 200 L to 3000 L—designed for the collection and storage of sterile process liquids such as cell culture media, process water or purification media. Options include temperature-control through integrated water jackets, BPC deploying hoists, as well as other options to optimize performance in your productions processes.
The Thermo Scientific HyPerforma Smartainer 3.0 BPC provides end-users producing biologics or vaccine therapeutics with a large-volume, sterile liquid storage system for volumes ranging from 200 L to 3000 L—designed for the collection and storage of sterile process liquids such as cell culture media, process water or purification media. Options include temperature-control through integrated water jackets, BPC deploying hoists, as well as other options to optimize performance in your productions processes.
TRUSTED_SUSTAINABILITY
Spécification
Capacité (métrique) 500 l
Product Type Bioprocess Container (BPC)
Type d’insert À soufflet vertical