Learn More
Thermo Scientific™ HyperSep™ SPE Plates
Codice prodotto. 30276730 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Comprend:
Plaque à 96 puits 10 mg/1 ml
Plaque à 96 puits 20 mg/1 ml
Plaque à 96 puits 3 mg/0.6 ml
Dimensione della confezione:
1 pièce
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Comprend Dimensione della confezione
30276730 Plaque à 96 puits 10 mg/1 ml 1 pièce
30276726 Plaque à 96 puits 20 mg/1 ml 1 pièce
30276728 Plaque à 96 puits 3 mg/0.6 ml 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 30276730 Fornitore Thermo Scientific™ N. del fornitore 6030010196WAX
 Article restreint
Expédition estimée: 30-11-2026
Aggiungi al carrello
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Innovative HyperSep™ Solid Phase Extraction Plates deliver clean, highly reproducible sample extracts with lower elution volumes—for greater sensitivity, reliability and cost savings.
Obtain clean, highly reproducible sample extracts with lower elution volumes using Thermo Scientific™ HyperSep™ Solid Phase Extraction Plates. The enhanced performance gives you greater confidence in your analytical results and lowers operating costs—without compromising ease of use or requiring complex method development. Thanks to an innovative and unique frit-less design, HyperSep SPE Well Plates deliver unparalleled performance, including:
  • Higher levels of reproducibility
  • Lower elution volumes
  • High extract cleanliness
HyperSep Solid Phase Extraction Plates achieve excellent recovery levels even with low volumes of extract solvents, resulting in a more concentrated analyte and increased sensitivity. You'll also realize additional cost and time saving from faster sample dry-down time and reduced solvent usage.
  • 10 mg/2 mL 96 well plate format
  • Hydrophobic Reversed Phase (HRP), Strong Cation Exchanger (SCX) and Strong Anion Exchanger (SAX) phase types
  • HyperSep products are also available in cartridge format.
TRUSTED_SUSTAINABILITY
Specifica
Poids du lit 10 mg
À utiliser avec (application) Bioanalytical Applications - Weak Ion-Exchange Retention of Strong Acidic Compounds. Sorbent Charge Can be Activated or Deactivated. Complementary Reversed Phase Retention of Neutral Compounds
Gamme de produits HyperSep
Type 96-well Plate
Volume (métrique) des puits 1 mL
Comprend Plaque à 96 puits 10 mg/1 ml
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.