Learn More
Enterobacter cloacae subsp. cloacae ATCC® 13047™†
Code produit 11926595
Encompass_Preferred
Change view
Click to view available options
Type de produit:
KWIK-STIK
Quantité:
2/paquet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Type de produit Quantité
11926595 KWIK-STIK 2/paquet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 11926595 Fournisseur Microbiologics Code fournisseur 0323P
For culture purposes and QC applications
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single Enterobacter cloacae subsp. cloacae strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2 or 6
KWIK-STIK™ Plus
  • Five self-contained KWIK-STIK units with microorganism limited to two passages from reference culture
  • LYFO DISK™
    • The most economical reference stock culture option; lyophilized pellet containing a singleEnterobacter cloacae subsp. cloacae strain
    • Easy to use; simply re-hydrate the pellet and inoculate
    • Available in a re-sealable vial containing 6 pellets
    • ATCC™ 13047™†
    †The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
    Spécification
    ATCC® 13047™*
    Niveau de sécurité biologique 1
    Certifications/Conformité CE, online certificate of analysis
    Contenu et stockage 2°C à 8°C
    Forme Lyophilized Pellet
    Type de milieu Kwik-stik™
    Type de produit KWIK-STIK
    À utiliser avec (application) Culture purpose and QC application
    Quantité 2/paquet
    Espèces Enterobacter cloacae subsp. cloacae