Learn More
MedChemExpress Indolicidin
Code produit 16411918
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16411918 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16411918 Fournisseur MedChemExpress Code fournisseur HYP0261
MedChemExpress Indolicidin is a potent antimicrobial peptide purified from the cytoplasmic granules of bovine neutrophils.
Spécification
CAS 140896-21-5
Contenu et stockage Protect from light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
À utiliser avec (application) Neuroscience-Neuromodulation
Poids moléculaire 1906.28 g/mol
Type de produit Natural Products
Quantité 5 mg
Conditions d’expédition Room temperature