Learn More
Ion 510™ & Ion 520™ & Ion 530™ Kit – Chef (1 sequencing run per initialization)
Shop All Thermo Scientific ProductsSpecifications
Specifications
| Software Type | Torrent Suite Software 5.6 |
| Quantité | 8 réactions |
| Gamme de produits | Ion Chef, Ion Torrent |
| Type de produit | Chef Kit |
| Conditions d’expédition | Approved for shipment at Room Temperature or on Wet Ice |
| Cadence | 400 Base-Read Libraries |
| Contenu et stockage | • Ion S5 Chef supplies, store at room temperature • Ion S5 Chef solutions, store at room temperature • Ion 510 and Ion 520 and Ion 530 Chef reagents, store at -5 to -30°C • Ion S5 Sequencing solutions, store at room temperature • Ion S5 Sequencing reagents, store at -5 to -30°C |
Usage exclusivement réservé à la recherche. Ne pas utiliser pour des procédures de diagnostic.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?