Learn More
Ion 510™ & Ion 520™ & Ion 530™ Kit – Chef (1 sequencing run per initialization)
Product Code. 18879226 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantité:
8 réactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité
18879226 8 réactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18879226 Supplier Thermo Scientific Supplier No. A34019
 Article restreint
Expédition estimée: 09-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Software Type Torrent Suite Software 5.6
Quantité 8 réactions
Gamme de produits Ion Chef, Ion Torrent
Type de produit Chef Kit
Conditions d’expédition Approved for shipment at Room Temperature or on Wet Ice
Cadence 400 Base-Read Libraries
Contenu et stockage • Ion S5 Chef supplies, store at room temperature
• Ion S5 Chef solutions, store at room temperature
• Ion 510 and Ion 520 and Ion 530 Chef reagents, store at -5 to -30°C
• Ion S5 Sequencing solutions, store at room temperature
• Ion S5 Sequencing reagents, store at -5 to -30°C