Learn More
Isoginkgetin, MedChemExpress
Code produit 16479258
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16479258 10 mg
16441678 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 16479258 Fournisseur Medchem Express Code fournisseur HYN2117
MedChemExpress Isoginkgetin is a pre-mRNA splicing inhibitor inhibitor. Isoginkgetin also inhibits activities of both Akt, NF-κB and MMP-9. Isoginkgetin inhibits the activity of the 20S proteasome, induces apoptosis and activates autophagy.
Isoginkgetin is a pre-mRNA splicing inhibitor inhibitor. Isoginkgetin also inhibits activities of both Akt, NF-κB and MMP-9. Isoginkgetin inhibits the activity of the 20S proteasome, induces apoptosis and activates autophagy.
Spécification
Nom chimique ou matériau Isoginkgetin
CAS 548-19-6
Couleur Light Yellow
Notes de qualité de la pureté Research
Formule moléculaire C32H22O10
Quantité 10 mg
Informations sur la solubilité DMSO : 100 mg/mL (176.52 mM; Need ultrasonic)
SMILES O=C1C=C(C2=CC=C(OC)C=C2)OC3=C(C4=CC(C5=CC(C6=C(O)C=C(O)C=C6O5)=O)=CC=C4OC)C(O)=CC(O)=C13
Poids moléculaire (g/mol) 566.51
Poids de la formule 566.51
Pourcentage de pureté 98.06%
Qualité Research
Forme physique Solid
À utiliser avec (application) Cancer-Kinase/protease