missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropanol D8 99% D, Eurisotop
Description
Isopropanol D8
- Isotopic Enrichment: 99.00% D
- Quantity: 10 g
- Water Content: water <0.1%
CoA and MSDS available at Eurisotop.com
Spécification
Spécification
| Nom chimique ou matériau | Isopropanol D8 |
| Couleur | Incolore |
| Deutérium (D) | 99.0% D |
| Quantité | 10 g |
| CAS | 22739-76-0 |
| Point de fusion | -89°C |
| Point d’ébullition | 83°C |
| Formule moléculaire | C3H8O |
| Synonyme | 2-Propanol-d8, Octadeuteroisopropanol, Isopropanol-d8 |
| Clé InChI | KFZMGEQAYNKOFK-PIODKIDGSA-N |
| Afficher plus |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?