Learn More
MedChemExpress L-Asparaginase
Code produit 16489668
Encompass_Preferred
Change view
Click to view available options
Quantité:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16489668 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16489668 Fournisseur MedChemExpress Code fournisseur HYP1923
MedChemExpress L-Asparaginase (L-ASNase) is a deamidating enzyme that catalyses the hydrolysis of L-asparagine and L-glutamine, and can be used for the research of acute lymphoblastic leukemia. L-Asparaginase depletes L-asparagine from plasma resulting in inhibition of RNA and DNA synthesis with the subsequent blastic cell apoptosis.
Spécification
Pureté 99.0%
CAS 9015-68-3
Contenu et stockage Protect from light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
À utiliser avec (application) Cancer-programmed cell death
Type de produit Peptides
Quantité 10 mg
Conditions d’expédition Room temperature
Solubilité 0.1 M HCL : 50 mg/mL (ultrasonic and adjust pH to 3 with HCl) ∣H2O : 1 mg/mL (ultrasonic and warming and heat to 50°C)
Synonyme L-​ASNase