Learn More
Thermo Scientific™ Lab Vision™ Citrate Buffer for Heat-Induced Epitope Retrieval (100X)
Code produit 12633947 Tous les produits Epredia Produits
Modifier l'affichage
Click to view available options
Quantité:
50 ml
Conditionnement:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
12633947 50 ml 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 12633947 Fournisseur Thermo Scientific™ Code fournisseur TA050CBX
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Voir les produits de remplacement
Les retours ne sont pas autorisés pour ce produit. View return policy
Improve the reactivity of many antibodies in formalin-fixed tissues with the Thermo Scientific™ Lab Vision™ Citrate Buffer for Heat-Induced Epitope Retrieval (100X).
Improve the reactivity of many antibodies in formalin-fixed tissues with the Thermo Scientific™ Lab Vision™ Citrate Buffer for Heat-Induced Epitope Retrieval (100X)
  • 1M Citrate buffer, pH 6.0
  • 100X stock solution
  • Diluted 100-fold with distilled water before use
Storage Conditions: Store at room temperature. Stable for up to 18 months. Contains no preservatives. For periods longer than 3 months, store at 2-8°C
TRUSTED_SUSTAINABILITY
Specifications
Capacité 50 mL
Contenu et stockage Température ambiante
Dilution 100X
Type de produit Citrate Buffer
Quantité 50 ml
pH 6
Utilisation réservée la recherche