Learn More
Lentinan (Technical Grade), TRC
Code produit 30479714
Change view
Click to view available options
Quantité:
5 g
10 g
25 g
Conditionnement:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30479714 10 g 10g
30469978 5 g 5g
30471456 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Code produit 30479714 Fournisseur Toronto Research Chemicals Code fournisseur TRCL32910010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 37339-90-5
Formule moléculaire (C42H70O35)nH2O
Poids moléculaire (g/mol) 1153
Synonyme Lentinan (Technical Grade)
Nom IUPAC (2R,3R,4S,5R,6R)-4-(((2S,3R,4S,5R,6R)-4-(((2S,3R,4S,5R,6R)-4-(((2S,3R,4S,5R,6R)-3,5-dihydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-6-((((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-3,5-dihydroxy-6-((((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-2,3,5-triol
SMILES O[C@H]([C@@H](CO)O[C@@H](O[C@H]1[C@H](O)[C@@H](CO[C@@H]([C@@H]2O)O[C@H](CO)[C@@H](O)[C@@H]2O)O[C@@H](O[C@H]3[C@H](O)[C@@H](CO)O[C@@H](O[C@H]4[C@H](O)[C@@H](CO[C@@H]([C@@H]5O)O[C@H](CO)[C@@H](O)[C@@H]5O)O[C@@H](O[C@H]6[C@H](O)[C@@H](CO)O[C@@H](O)[C@@H]6O)[C@@H]4O)[C@@H]3O)[C@@H]1O)[C@@H]7O)[C@@H]7O[H]
Spécification
Nom chimique ou matériau Lentinan
Quantité 10 g
Formule InChI InChI=1S/C42H72O36/c43-1-8-15(48)22(55)25(58)37(69-8)66-6-13-20(53)34(76-39-27(60)24(57)17(50)10(3-45)71-39)30(63)42(74-13)77-33-19(52)12(5-47)72-40(29(33)62)78-35-21(54)14(7-67-38-26(59)23(56)16(49)9(2-44)70-38)73-41(31(35)64)75-32-18(51)11(4-46)68-36(65)28(32)61/h8-65H,1-7H2/t8-,9-,10-,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22+,23+,24+,25-,26-,27-,28-,29-,30-,31-,32+,33+,34+,35+,36-,37-,38-,39+,40+,41+,42+/m1/s1