Description
When adjusted for molarity, all three salt forms are equally effective; however, the hydrochloride salt is usually less invasive in biological settings.
Inhibits trypsin-like proteases and some cysteine proteases including: endoproteinase Lys-C, kallikrein, papain, thrombin, cathepsin B, cathepsins H and L, trypsin, calpain, trypsin8 and plasmin.
Spécification
Spécification
| CAS | 103476-89-7 |
| Contenu et stockage | Store at −20°C, desiccate, protect from light. |
| Format | White Powder |
| Poids moléculaire | 427.57 |
| Quantité | 100 mg |