Learn More
Invitrogen™ iBlot™ 2 Gel Horizontal Transfer Device
Performs Western blotting transfer simply, efficiently, and reliably, within seven minutes and without the need for liquid buffers
Marque:  Invitrogen™ IB21001PR
Règlementaire : Cet article est soumis à éco-contribution.
Code produit. 15502897
  • 2215.00€ / 1 pièces
Requires : iBlot 2 device and transfer stacks
Description
Description
The iBlot 2 Gel Transfer Device is an integral part of the iBlot 2 Dry Blotting System, which consists of the transfer device and consumable transfer stacks that contain the required buffers and transfer membrane (nitrocellulose or PVDF).
  • Complete protein transfer in 7 minutes or less
  • High detection sensitivity and even transfer
  • Increased blotting reliability and reproducibility
  • Flexible gel size formats and membrane types
  • A simple, user-friendly system
  • Options to create new custom programs
  • Built-in tutorial and application notes
  • High-quality transfer stacks that are more compact than before
Rapid, High-quality Protein Transfers
  • For dry electroblotting of proteins from mini-, midi-, and E-PAGE™ gels onto nitrocellulose or PVDF membranes for Western detection
  • Offers high-quality transfer, convenience, and speed, producing crisp, clear bands that remain sharp and straight, with exceptional transfer efficiency
  • Allows users to create custom programs
iBlot 2 Transfers Save Time
  • With the iBlot 2 system, there is no need to prepare buffers, pretreat the gel, or clean up after blotting
  • The total preparation and run time is normally less than 10 minutes per blot for significant time savings compared to conventional Western transfer techniques
How It Works
  • Buffer ion reservoirs are incorporated into the gel matrix (transfer stacks) instead of buffer tanks or soaked papers
  • The high density of ions in the gel matrix enables rapid protein transfer
  • During blotting, the copper anode does not generate oxygen gas as a result of water electrolysis, reducing blot distortion (conventional protein transfer techniques, including wet, semi-wet, and semi-dry, use inert electrodes that generate oxygen)
  • Transfer time is reduced by the shortened distance between electrodes, high field strength, and high current
  • Trapped air bubbles, often created during the manual preparation of the blotting sandwich layers, are easy to avoid due to the de-bubbling design that promotes even and complete transfer
Proteins, Expression, Isolation and Analysis, Western Blotting
For Research Use Only. Not for use in diagnostic procedures.