Learn More
Phenomenex™ Luna™ 5 μm Phenyl-Hexyl 100 Å, LC Column 150 x 4.6 mm
Description
- The Luna™ brand of columns and media is more than just a product line from Phenomenex
- It is a pledge to provide you with the highest level of satisfaction for your chromatographic goals
- The Luna™ Phenyl Hexyl is our most hydrophobic phenyl column and it will also provide good hydrogen accepting functionality for acidic retention
Spécification
Spécification
| Diamètre (métrique) | 4.6 mm |
| Longueur (métrique) | 150 mm |
| Granulométrie | 5 μm |
| pH | 1.5 to 9 |
| Porosité | 100 Å |
| Taux de carbone | 17.5% |
| Matériau | Silica |
| Technique | HPLC, LC/MS, UHPLC, UHPLC-MS |
| Type de produit | HPLC Column |