missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lycopersicon Esculentum (Tomato) Lectin (LEL, TL), Texas Red
Description
- Lycopersicon esculentum (Tomato) Lectin (LEL, TL), Texas Red, is a bright-red LEL-fluorophore conjugate. The tomato lectin LEL is widely used as marker of blood vessels and microglial cells in rodents and in neuroscience research.
- It is a valuable tool in studies of rodent tumor angiogenesis and the tracing of neovascular development in xenograft models and brain research.
- LEL-Texas Red can be applied directly to tissue sections, or it can be introduced through intravascular perfusion for fluorescence detection.
- Very bright Texas Red conjugate detected with traditional FITC filter set, Ex/Em: 595/615 nm
- Applications: immunofluorescence, glycobiology
- Sugar specificity: [GlcNAc] 1,3-N-acetylglucosamine
- Inhibiting/eluting sugar: chitin hydrolysate
- Blood group specificity: non-specific
- Unconjugated lectin molecular weight: 71 KDa.
- Immunofluorescence working concentration: 5–20 μg/mL
- LEL is a very stable single-subunit glycoprotein containing ∼50 percent arabinose and galactose and may form multimeric aggregates in solution.
- It is provided at a 1 mg/mL concentration in 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide, 0.1 mM Ca2+. If a precipitate forms upon long-term storage, warm to 37°C.
Spécification
Spécification
| Quantité | 1 mg |
| Source | Tomato |
| Conjugué | Texas Red |
| Contenu et stockage | When stored at or below 2–8°C in the dark, the product is stable for 6 months after reciept. |
| Forme | Solution |
| Sous-type de protéine | Lectins |
For Research Use Only. Not for use in diagnostic procedures.
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?