Description
Use our tool to find and order pre-designed primers for PCR and Sanger sequencing. Free of SNPs and dimers, target-specific, and suitable for universal PCR conditions, these primers can be ordered unmodified, M13-tailed, HPLC-purified, or desalted. All primers are checked by mass spectrometry.
Specifications
Specifications
| Description | Amorce sens humaine HPLC M13, 50 nmole, pour le séquençage de PCR et de Sanger |
| Type de produit | Amorce sens humaine HPLC M13 |
| Primer | Amorce sens |
| Quantité | 50 nmol |
| Qualité | HPLC |