Learn More
Madrasin, MedChemExpress
Code produit 16482546
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
10 mg
25 mg
50 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16482546 25 mg
16469516 50 mg
16455607 5 mg
16432868 10 mg
16404276 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Code produit 16482546 Fournisseur Medchem Express Code fournisseur HY100236
MedChemExpress Madrasin (DDD00107587) is a splicing inhibitor that prevents formation of both splicing intermediates and products in vitro and interferes with one or more early steps in the pathway of spliceosome assembly. Madrasin also can inhibit pre-mRNA splicing in vitro and modify splicing of endogenous pre-mRNA in cells.
Madrasin (DDD00107587) is a splicing inhibitor that prevents formation of both splicing intermediates and products in vitro and interferes with one or more early steps in the pathway of spliceosome assembly. Madrasin also can inhibit pre-mRNA splicing in vitro and modify splicing of endogenous pre-mRNA in cells.
Spécification
Nom chimique ou matériau Madrasin
CAS 374913-63-0
Couleur Blanc
Notes de qualité de la pureté Research
Formule moléculaire C16H17N5O2
Quantité 25 mg
Synonyme DDD00107587
Informations sur la solubilité DMSO : 2 mg/mL (6.42 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C1N=C(NC2=NC(C)=C3C=CC(OC)=CC3=N2)NC(C)=C1C
Poids moléculaire (g/mol) 311.34
Poids de la formule 311.34
Pourcentage de pureté 99.42%
Qualité Research
Forme physique Solid
À utiliser avec (application) Cancer-programmed cell death