Learn More
Invitrogen™ MAGPIX™ Drive Fluid Plus
Code produit 30245082
Modifier l'affichage
Click to view available options
Quantité:
4 x 700 ml
Conditionnement:
4 pièces
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
30245082 4 x 700 ml 4 pièces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 30245082 Fournisseur Invitrogen™ Code fournisseur 4050030
 Article restreint
En stock
Ajouter au panier
Edge
Ajouter au panier
MAGPIX Drive Fluid Plus is the delivery medium for transporting the sample being analyzed on the Luminex MAGPIX system to the optic components.
MAGPIX Drive Fluid Plus is the delivery medium for transporting the sample being analyzed on the Luminex MAGPIX system to the optic components. Each unit in the 4-pack of drive fluid contains 700 mL.
Please refer to the Luminex instrument user manual for proper installation and use of MAGPIX Drive Fluid Plus with the MAGPIX System.
This reformulated sheath product replaces the original MAGPIX Drive Fluid (Cat. No. MPXDF4PK). The new sheath formulation is updated from a single preservative to a mixture of three preservatives to allow for greater transit stability and to reduce the likelihood of future acquired microbial resistance.
Usage exclusivement réservé à la recherche. Ne pas utiliser pour des procédures de diagnostic. Non utilisé à des fins thérapeutiques ou diagnostiques humaines ou animales.
TRUSTED_SUSTAINABILITY
Spécification
Gamme de produits MAGPIX
Quantité 4 x 700 ml