Learn More
Mal-amido-peg6-NHS, TRC
Code produit 30472245
Change view
Click to view available options
Quantité:
10 mg
50 mg
100 mg
Conditionnement:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30472245 50 mg 50mg
30469110 10 mg 10mg
30470006 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Code produit 30472245 Fournisseur Toronto Research Chemicals Code fournisseur TRCM11105350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 1137109-21-7
Formule moléculaire C26H39N3O13
Synonyme Maleimide-PEG6-succinimidyl Ester∣N-Hydroxysuccinimidyl-(polyethylene glycol) 6-Maleimide∣SM(PEG)6
Spécification
Certifications/Conformité ISO 9001
Poids moléculaire 601.6
Conditions d’expédition Room Temperature