Learn More
MedChemExpress Mastoparan
Code produit 16414016
Encompass_Preferred
Change view
Click to view available options
Quantité:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16414016 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16414016 Fournisseur MedChemExpress Code fournisseur HYP0246
MedChemExpress Mastoparan, a tetradecapeptide which is a component of wasp venom, stimulates release of prolactin from cultured rat anterior pituitary cells.
Spécification
Pureté 95.47%
CAS 72093-21-1
Contenu et stockage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
À utiliser avec (application) Metabolism-protein/nucleotide metabolism
Poids moléculaire 1478.91 g/mol
Type de produit Natural Products
Quantité 25 mg
Conditions d’expédition Room temperature
Solubilité H2O : 33.33 mg/mL (22.54 mM∣Need ultrasonic)