Learn More
MedChemExpress Mastoparan
Code produit 16492709
Encompass_Preferred
Change view
Click to view available options
Quantité:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16492709 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16492709 Fournisseur MedChemExpress Code fournisseur HYP0246
MedChemExpress Mastoparan, a tetradecapeptide which is a component of wasp venom, stimulates release of prolactin from cultured rat anterior pituitary cells.
Spécification
Pureté 95.47%
CAS 72093-21-1
Contenu et stockage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
À utiliser avec (application) Metabolism-protein/nucleotide metabolism
Poids moléculaire 1478.91 g/mol
Type de produit Natural Products
Quantité 1 mg
Conditions d’expédition Room temperature
Solubilité H2O : 33.33 mg/mL (22.54 mM∣Need ultrasonic)