Learn More
Mepenzolate Bromide, TRC
Code produit 30482021
Change view
Click to view available options
Quantité:
100 mg
1 g
5 g
Conditionnement:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30482021 5 g 5g
30472545 1 g 1g
30475504 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Code produit 30482021 Fournisseur Toronto Research Chemicals Code fournisseur TRCM2237505G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 76-90-4
Formule moléculaire C21 H26 N O3 . Br
Poids moléculaire (g/mol) 420.34
Synonyme Piperidinium, 3-[(2-hydroxy-2,2-diphenylacetyl)oxy]-1,1-dimethyl-, bromide (1:1), 3-Hydroxy-1,1-dimethylpiperidinium bromide benzilate (6CI,7CI), Piperidinium, 3-[(hydroxydiphenylacetyl)oxy]-1,1-dimethyl-, bromide (9CI), Piperidinium, 3-hydroxy-1,1-dimethyl-, bromide, benzilate (8CI), Benzilic acid, ester with 3-hydroxy-1,1-dimethylpiperidinium bromide (8CI), 1-Methyl-3-piperidyl benzilate methylbromide, Cantil, Cantilaque, Cantilon, Cantril, Colibantil, Colopiril, Colum, Delevil, Gastropidil, JB 340, Mepenzolate bromide, N-Methyl-3-piperidyl benzilate methyl bromide, N-Methyl-3-piperidyldiphenylglycolate methobromide, NSC 4358, Trancolon
Nom IUPAC (1,1-dimethylpiperidin-1-ium-3-yl) 2-hydroxy-2,2-diphenylacetate;bromide
SMILES [Br-].C[N+]1(C)CCCC(C1)OC(=O)C(O)(c2ccccc2)c3ccccc3
Spécification
Nom chimique ou matériau Mepenzolate Bromide
Quantité 5 g
Formule InChI InChI=1S/C21H26NO3.BrH/c1-22(2)15-9-14-19(16-22)25-20(23)21(24,17-10-5-3-6-11-17)18-12-7-4-8-13-18;/h3-8,10-13,19,24H,9,14-16H2,1-2H3;1H/q+1;/p-1
Poids de la formule 419.1096