Learn More
Methenamine Hippurate, TRC
Code produit 30479195
Change view
Click to view available options
Quantité:
1 g
10 g
Conditionnement:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30479195 1 g 1g
30475942 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 30479195 Fournisseur Toronto Research Chemicals Code fournisseur TRCM2260251G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 5714-73-8
Formule moléculaire C9 H9 N O3 . C6 H12 N4
Poids moléculaire (g/mol) 319.36
Synonyme Glycine, N-benzoyl-, compd. with 1,3,5,7-tetraazatricyclo[3.3.1.13,7]decane (1:1), Hippuric acid, compd. with hexamethylenetetramine (1:1) (8CI), 1,3,5,7-Tetraazatricyclo[3.3.1.13,7]decane, compd. with N-benzoylglycine (1:1), Hexamethylenetetramine, monohippurate (8CI), Haiprex, Hexamethylenetetramine hippurate, Hexamethylenetetramine hippuric acid complex, Hexamine hippurate, Hippramine, Hiprex, Methenamine hippurate, R-657, Urex, Viapta
SMILES OC(=O)CNC(=O)c1ccccc1.C2N3CN4CN(CN2C4)C3
Specifications
Nom chimique ou matériau Methenamine Hippurate
Quantité 1 g
Formule InChI InChI=1S/C9H9NO3.C6H12N4/c11-8(12)6-10-9(13)7-4-2-1-3-5-7;1-7-2-9-4-8(1)5-10(3-7)6-9/h1-5H,6H2,(H,10,13)(H,11,12);1-6H2
Poids de la formule 319.1644