Learn More
Microbiologics™ Clostridium sporogenes ATCC® 19404™† Epower™
Code produit 11976575
Encompass_Preferred
Modifier l'affichage
Click to view available options
Espèces:
Clostridium sporogenes
Conditionnement:
1 pièce
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Espèces Conditionnement
11976575 Clostridium sporogenes 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 11976575 Fournisseur Microbiologics™ Code fournisseur 317000
 Article restreint
Cet article nécessite une mise à jour du prix avant d'être commandé.
Demandez un prix pour cet article ici.
missing translation for 'validMainframeMsg'
For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination
Epower
  • Lyophilized microorganism preparations
  • Biosafety Level: 1
  • Pellets can be used individually or combined for a mixed microorganism population challenge
  • Easily manipulated to deliver a wide variety of concentrations
  • Available in concentrations ranging from 102 to 108 CFU per pellet
  • ATCC™ 19404™†
Spécification
ATCC® ATCC 19404
Forme Pellets
Type de produit QC Microorganism
Quantité 10/Pk.
Espèces Clostridium sporogenes
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.