Learn More
Microbiologics™ X10 DISQUE K. RHIZOPHILA ATCC 9341
Code produit 11981456
Encompass_Preferred
Modifier l'affichage
Click to view available options
Espèces:
Kocuria rhizophila
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Espèces
11981456 Kocuria rhizophila
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 11981456 Fournisseur Microbiologics™ Code fournisseur 0688E3
 Article restreint
Expédition estimée: 02-12-2026
Ajouter au panier
Ajouter au panier
missing translation for 'validMainframeMsg'
Includes
Product warranty and peel-off certificate of assay
Lyophilized pellets add convenience to special projects and internal proficiency plans
Informations sur la commande
Available in vials containing 10 lyophilized pellets, with a range of concentration from 102 to 109 per pellet
Spécification
ATCC® 9341
Niveau de sécurité biologique 1
Forme Lyophilized Pellet
Type de produit QC Microorganism
À utiliser avec (application) Lyophilized pellets add convenience to special projects and internal proficiency plans.
Comprend Product warranty and peel-off certificate of assay
Valeur de dosage moyenne 1.0-9.9E+03 CFU per pellet
Type d’emballage Vial
Quantité 10/Pk.
Espèces Kocuria rhizophila
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.