Learn More
Microbiologics™ Préparations de microorganismes EZ-Accu Shot™
Código de producto. 11966805
Encompass_Preferred
Cambiar vista
Click to view available options
Espèces:
Aspergillus brasiliensis
Bacillus subtilis
Clostridium sporogenes
Escherichia coli
Pseudomonas aeruginosa
Staphylococcus aureus
ATCC®:
11437™*
6538™*
8739™*
9027™*
ATCC 16404
ATCC 19404
ATCC 6633
Tamaño de la unidad:
1 pièce
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Espèces ATCC® unitSize
11966805 Clostridium sporogenes ATCC 19404 -
11936775 Aspergillus brasiliensis ATCC 16404 1 pièce
11966775 Escherichia coli 8739™* 1 pièce
11986805 Pseudomonas aeruginosa 9027™* 1 pièce
11996805 Staphylococcus aureus 6538™* 1 pièce
11946775 Bacillus subtilis ATCC 6633 1 pièce
11976805 Clostridium sporogenes 11437™* 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11966805 Proveedor Microbiologics™ N.º de proveedor 0317A
 Article restreint
Expédition estimée: 17-08-2026
Añadir a la cesta
Añadir a la cesta
Este artículo no se puede devolver. Vea la política de devoluciones
Includes
Product warranty and peel-off Certificate of Assay.
For easy growth promotion testing of culture media
Each lyophilized microorganism preparation is designed to deliver less than 100CFU per 0.1mL of hydrated suspension as recommended by the USP/JP/Ph. Eur. With little to no prep time required and up to eight hours of stability, EZ-Accu Shot adds efficiency and flexibility to QC lab schedules. Each 1.2mL vial of hydrated suspension offers 10 inocula. For added convenience, a peel-off Certificate of Assay is included for quality control documentation.
  • Instant dissolve, no pre-incubation for minimal prep time
  • One pellet per vial eliminates the need for forceps, reducing the risk of contamination
  • Eight-hour stability extends time and flexibility to perform tests
  • Precise CFU count ensures accurate results
  • Easy storage with no dry ice required, reducing shipping costs
  • Peel-off Certificate of Assay for complete, convenient documentation
  • Online Certificate of Analysis for documentation of phenotypic, macroscopic and microscopic test results
  • Traceable to USP reference culture or assured quality
Información de órdenes
Each kit contains five vials of a single enumerated microorganism (one lyophilized pellet per vial) and five vials of hydrating fluid (1.2mL per vial).
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Especificaciones
ATCC® ATCC 19404
Forme Pellets
Type de produit EZ-Accu Shot
Espèces Clostridium sporogenes
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.