Learn More
Microbiologics™ X10 DISQUE BACIL SUBTILIS bacillus subtilis ATCC® 6633 (tm) ez-cfu/1000 à
Code produit 11901426
Encompass_Preferred
Modifier l'affichage
Click to view available options
Espèces:
B. subtilis sous-espèce spizizenni
Conditionnement:
10 pièces
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Espèces Conditionnement
11901426 B. subtilis sous-espèce spizizenni 10 pièces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 11901426 Fournisseur Microbiologics™ Code fournisseur 0486C
 Article restreint
Expédition estimée: 15-12-2026
Ajouter au panier
Ajouter au panier
missing translation for 'validMainframeMsg'
Includes
Product warranty and peel-off certificate of assay
Assayed microorganism preparation for USP quantitative microorganism challenges
Lyophilized microorganism preparations delivering the desired 10 to 100CFU/0.1mL of challenge material (following hydration and dilution of 1.0mL of hydrated suspension in 9.0mL of pH 7.2 phosphate buffer).
  • Traceable to reference cultures
Informations sur la commande
Supplied as two vials of a single assayed microorganism (10 lyophilized pellets in each vial) and 10 vials of hydrating fluid (2.0mL in each vial)
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.
Spécification
ATCC® 6633
Format EZ-CFU
Type de produit EZ-CFU
À utiliser avec (application) Assayed microorganism preparations for USP quantitative microorganism challenges.
Comprend Product warranty and peel-off certificate of assay
Type d’emballage Vial
Espèces B. subtilis sous-espèce spizizenni