Learn More
Microbiologics™ EZ-CFU S. SEROVAR TYPHIMURIUM ATCC® 13311
Code produit 14802321
Encompass_Preferred
Modifier l'affichage
Click to view available options
Espèces:
Salmonella enterica sous-espèce enterica sérovar typhimurium
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Espèces
14802321 Salmonella enterica sous-espèce enterica sérovar typhimurium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 14802321 Fournisseur Microbiologics™ Code fournisseur 0421C
 Article restreint
Expédition estimée: 02-12-2026
Ajouter au panier
Ajouter au panier
missing translation for 'validMainframeMsg'
Includes
Product warranty and peel-off certificate of assay
Assayed microorganism preparation for USP quantitative microorganism challenges
Lyophilized microorganism preparations delivering the desired 10 to 100CFU/0.1mL of challenge material (following hydration and dilution of 1.0mL of hydrated suspension in 9.0mL of pH 7.2 phosphate buffer).
  • Traceable to reference cultures
Informations sur la commande
Supplied as two vials of a single assayed microorganism (10 lyophilized pellets in each vial) and 10 vials of hydrating fluid (2.0mL in each vial)
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.
Spécification
ATCC® 13311
Niveau de sécurité biologique 2
Format EZ-CFU
Type de produit EZ-CFU
À utiliser avec (application) Assayed microorganism preparations for USP quantitative microorganism challenges.
Comprend Product warranty and peel-off certificate of assay
Type d’emballage Vial
Espèces Salmonella enterica sous-espèce enterica sérovar typhimurium