Learn More
Microbiologics™ X20 DISQUES S.ENTERICA ATCC 14028 EZ-CFU
Beschreibung
Beinhaltet
Product warranty and peel-off certificate of assay
Lyophilized microorganism preparations delivering the desired 10 to 100CFU/0.1mL of challenge material (following hydration and dilution of 1.0mL of hydrated suspension in 9.0mL of pH 7.2 phosphate buffer).
- Traceable to reference cultures
Bestellinformation
Supplied as two vials of a single assayed microorganism (10 lyophilized pellets in each vial) and 10 vials of hydrating fluid (2.0mL in each vial)
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.
Spezifikation
Spezifikation
| ATCC® | 14028 |
| Niveau de sécurité biologique | 2 |
| Format | EZ-CFU |
| Type de produit | EZ-CFU |
| À utiliser avec (application) | Assayed microorganism preparations for USP quantitative microorganism challenges. |
| Comprend | Product warranty and peel-off certificate of assay |
| Type d’emballage | Vial |
| Espèces | Salmonella enterica sous-espèce enterica sérovar typhimurium |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Datenschutzrichtlinie.