Learn More
Microbiologics™ Bacillus cereus ATCC® 11778™†
Code produit 11936565
Encompass_Preferred
Modifier l'affichage
Click to view available options
Type de milieu:
Kwik-stik™
Quantité:
2/paquet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Type de milieu Quantité
11936565 Kwik-stik™ 2/paquet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 11936565 Fournisseur Microbiologics™ Code fournisseur 0256P
Please to purchase this item. Need a web account? Register with us today!
For culture purposes and QC applications
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single Bacillus cereus strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2 or 6
LYFO DISK™
  • The most economical reference stock culture option; lyophilized pellet containing a single Bacillus cereus strain
  • Easy to use; simply re-hydrate the pellet and inoculate
  • Available in a re-sealable vial containing 6 pellets
  • ATCC™ 11778™†
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Spécification
ATCC® 11778™*
Niveau de sécurité biologique 1
Certifications/Conformité CE, online certificate of analysis
Contenu et stockage 2°C à 8°C
Forme Lyophilized Pellet
Type de milieu Kwik-stik™
Type de produit Quality Control Organism
À utiliser avec (application) Culture purpose and QC application
Quantité 2/paquet
Espèces Bacillus cereus