Learn More
Microbiologics™ Corynebacterium urealyticum ATCC® 43044™†
Code produit 13487376
Encompass_Preferred
Modifier l'affichage
Click to view available options
Type de produit:
KWIK-STIK
Quantité:
2/paquet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Type de produit Quantité
13487376 KWIK-STIK 2/paquet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 13487376 Fournisseur Microbiologics™ Code fournisseur 01039P
 Article restreint
Expédition estimée: 15-12-2026
Ajouter au panier
Ajouter au panier
missing translation for 'validMainframeMsg'
For culture purposes and QC applications
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single Corynebacterium urealyticum strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2 or 6
LYFO DISK™
  • The most economical reference stock culture option; lyophilized pellet containing a single Corynebacterium urealyticum strain
  • Easy to use; simply re-hydrate the pellet and inoculate
  • Available in a re-sealable vial containing 6 pellets
  • ATCC™ 43044™†
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Spécification
ATCC® 43044
Niveau de sécurité biologique 2
Type de produit KWIK-STIK
Quantité 2/paquet
Espèces Corynebacterium urealyticum