Learn More
MedChemExpress Mifamurtide
Code produit 16455808
Encompass_Preferred
Change view
Click to view available options
Quantité:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16455808 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16455808 Fournisseur MedChemExpress Code fournisseur HY13682
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Mifamurtide (MTP-PE), an analog of the muramyl dipeptide (MDP), is a nonspecific immunomodulator by stimulating the immune response activating macrophages and monocytes. Mifamurtide, an orphan drug, is a specific ligand of NOD2 used as an insulin sensitizer. Mifamurtide has the potential for osteosarcoma research.
Spécification
Pureté 98.0%
CAS 83461-56-7
Contenu et stockage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
À utiliser avec (application) Cancer-programmed cell death
Poids moléculaire 1237.5 g/mol
Type de produit Reference compound
Quantité 10 mg
Conditions d’expédition Room temperature
Solubilité DMSO : 100 mg/mL (80.81 mM∣Need ultrasonic)
Synonyme MTP-PE∣ L-MTP-PE∣ CGP 19835