Learn More
MedChemExpress MLCK inhibitor peptide 18
Code produit 16442809
Encompass_Preferred
Change view
Click to view available options
Quantité:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16442809 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16442809 Fournisseur MedChemExpress Code fournisseur HYP1029
MedChemExpress MLCK inhibitor peptide 18 is a myosin light chain kinase (MLCK) inhibitor with an IC50 of 50 nM, and inhibits CaM kinase II only at 4000-fold higher concentrations.
Spécification
Pureté 99.66%
CAS 224579-74-2
Contenu et stockage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
Poids moléculaire 1324.62 g/mol
Type de produit Reference compound
Quantité 1 mg
Conditions d’expédition Room temperature
Solubilité H2O : 100 mg/mL (75.49 mM∣Need ultrasonic)