Learn More
MedChemExpress MLCK inhibitor peptide 18
Code produit 16444046
Encompass_Preferred
Change view
Click to view available options
Quantité:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16444046 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16444046 Fournisseur MedChemExpress Code fournisseur HYP1029
MedChemExpress MLCK inhibitor peptide 18 is a myosin light chain kinase (MLCK) inhibitor with an IC50 of 50 nM, and inhibits CaM kinase II only at 4000-fold higher concentrations.
Spécification
Pureté 99.66%
CAS 224579-74-2
Contenu et stockage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
Poids moléculaire 1324.62 g/mol
Type de produit Reference compound
Quantité 25 mg
Conditions d’expédition Room temperature
Solubilité H2O : 100 mg/mL (75.49 mM∣Need ultrasonic)