Learn More
R&D Systems™ Mouse CXADR (NP_001020363) VersaClone cDNA
Description
The Mouse CXADR (NP_001020363) VersaClone cDNA clone contains the complete ORF for CXADR, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The CXADR insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Spécification
Spécification
| Expression | mCxadr |
| Numéro d’adhésion | NP_001020363 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formule | cDNA is provided in 10 mM Tris-Cl |
| Type d’insert | 1111bp |
| pH | 8.5 |
| Identification génétique (Entrez) | 1525 |
| Contenu et stockage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Espèces | Mouse |
| Quantité | 10 μg |