Learn More
R&D Systems™ Mouse Glut5 (NP_062715) VersaClone cDNA
Code produit 18757178
Modifier l'affichage
Click to view available options
Quantité:
10 μg
Conditionnement:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
18757178 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 18757178 Fournisseur R&D Systems™ Code fournisseur RDC1021
  • ONLINE EXCLUSIVE PRICE
    341.00€
    322.35€ / 10µg
    Économisez 18.65€
    5% de réduction
 Article restreint
Expédition estimée: 19-10-2026
Solute carrier family 2 (facilitated glucose transporter), member 5
The Mouse Glut5 (NP_062715) VersaClone cDNA clone contains the complete ORF for Glut1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Glut1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Informations sur la commande
Ships at ambient temperature.
TRUSTED_SUSTAINABILITY
Spécification
Alias de gène Choreoathetosis/Spasticity, Episodic (Paroxysmal Choreoathetosis), CSE, DYT17, DYT18, DYT9, EIG12, Glucose transporter type 1, erythrocyte/brain, GLUT-1, GLUT1DS, HepG2 glucose transporter, HTLVR, Human T-Cell Leukemia Virus (I and II) Receptor, MGC141895, MGC141896, PED, SLC2A1, solute carrier family 2 (facilitated glucose transporter), member 1, Solute Carrier Family 2 Member 1, solute carrier family 2, facilitated glucose transporter member 1
Contenu et stockage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantité 10 μg
For Reasearch Use Only; Not for Use in Humans