missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IFN-gamma (ELISA Standard) Recombinant Protein

Code produit 15578906
Change view
Click to view available options
Quantité:
4 Paquets
Conditionnement:
4 pièces
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
15578906 4 Paquets 4 pièces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 15578906 Fournisseur Invitrogen™ Code fournisseur 39831165

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Recombinant Protein

Description: Mouse IFN-gamma is a ~20 kDa factor produced by activated T, B and NK cells, and is an anti-viral and anti-parasitic cytokine. IFN-gamma, in synergy with other cytokines such as TNF-alpha, inhibits proliferation of normal and transformed cells. Immunomodulatory effects of IFN-gamma are exerted on a wide range of cell types expressing the high affinity receptors for IFN-gamma. Glycosylation of IFN-gamma does not affect its biological activity. Applications Reported: Recombinant mouse IFN gamma is biologically active. The intended use of the Recombinant Mouse IFN gamma Single-Use ELISA RSG Standard is as a standard for mouse IFN gamma Ready-SET-Go! ELISA and mouse IFN gamma matched antibody pair ELISA. Typically the standard is run in a series of 7 doubling dilutions from the 'top' standard described below. Instructions for use (XMG1.2/R4-6A2 assays): For Ready-SET-Go! ELISA Set (TMB substrate). Dilution: Add 20 μL of standard solution to 10 mL of assay diluent to prepare the top standard solution (2000 pg/mL).For matched antibody pair ELISA (ABTS or Super AquaBlue substrate). Dilution: Add 20 μL of standard solution to 10 mL of assay diluent to prepare the top standard solution (2000 pg/mL). Instructions for use (AN-18/R4-6A2 assays): For Ready-SET-Go! ELISA Set (TMB substrate). Dilution: Add 20 μL of standard solution to 10 mL of assay diluent to prepare 2000 pg/mL solution. Mix thoroughly.

IFN gamma (Interferon gamma, Type II interferon) is a macrophage activation factor, and immune interferon that is produced primarily by T-lymphocytes and natural killer cells in response to antigens, mitogens, Staphylococcus enterotoxin B, phytohemaglutanin and other cytokines. IFN gamma is a dimeric protein consisting of two 146 amino acid subunits. IFN gamma is a glycoprotein that exists, functionally, as a homodimer of approximately 45 kDa. On SDS-PAGE, IFN gamma appears as a combination of 25, 20 and minor 15.5 kDa bands as a result of differential glycosylation. The biological activity of the IFN gamma homodimer is highly species specific. Human IFN gamma does not show cross-reactivity with mouse. IFN gamma function includes the following: antiviral activity, tumor antiproliferative activity, induction of class I and II MHC, macrophage activation, and enhanced immunoglobulin secretion by B lymphocytes. IFN gamma is involved in cytokine regulation and also acts synergistically with other cytokines. Activation of IFN gamma takes place through binding of IFN gamma receptor I and II, and activating the JAK-STAT pathway. IFN gamma does not show any homology with IFN alpha or IFN beta but human IFN gamma shows about 40% sequence homology with mouse IFN gamma. IFN gamma is upregulated by IL2, FGF basic, EGF and downregulated by vitamin D3 or DMN. IFN gamma gene mutations are associated with aplastic anemia.
TRUSTED_SUSTAINABILITY

Spécification

Numéro d’adhésion P01580
À utiliser avec (application) Neutralization, ELISA Standard
Formule Protein with 0.09% sodium azide
Identification génétique (Entrez) 15978
Poids moléculaire 20 kDa
Nom Mouse IFN-gamma (ELISA Standard)
Méthode de purification Purified
Quantité 4 Paquets
Source E. Coli
Conditions de stockage 2-8°C
État réglementaire RUO
Alias de gène BoIFNG; cytokine; EGK_03901; gamma interferon; gamma interferon precursor; gamma-interferon; gamma-interferon precursor (AA -23 to 143); H-IFN-g; IFG; IFI; IFN gamma; IFN γ; Ifng; IFN-g; IFNG2; IFN-gamma; IFN-gamma precursor; IFN-y; IFNγ; Immune interferon; Interferon; interferon gamma; interferon gamma precursor; interferon gamma type 2; Interferon y; Interferon γ; interferon, gamma; interferon-gama; interferon-gamma; Interferon-gamma level; interferon-gamma precursor; Interferonγ; M-IFN-g; R-IFN-g
Nom usuel IFN gamma
Symbole de gène(s) IFNG
Type de produit Protein
Conjugué Unconjugated
Espèces E. coli
Recombinant Recombinant
Séquence Mouse IFN-gamma, amino acids His23-Cys155 (Accession #NM_008337)
Contenu et stockage -80°C
Système d’expression E. coli
Forme Lyophilized
Sous-type de protéine Recombinant
Pureté ou qualité >98% by SDS-PAGE
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.