Learn More
R&D Systems™ Mouse IL-10 R alpha (NP_032374) VersaClone cDNA
Description
This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.
Informations sur la commande
Ships at ambient temperature
Spécification
Spécification
| Expression | IL-10 R alpha |
| Numéro d’adhésion | NP_032374 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formule | cDNA is provided in 10mM Tris-Cl, pH 8.200 |
| Type d’insert | 1741bp |
| Identification génétique (Entrez) | 3587 |
| Alias de gène | CD210 antigen, CDw210a, HIL-10R, IBD28, IL-10 receptor subunit alpha, IL10R alpha, IL-10R subunit 1, IL-10R1, IL10RA, IL-10RA, IL10RIL-10R subunit alpha, interleukin 10 receptor, alpha, interleukin-10 receptor alpha chain, Interleukin-10 receptor subunit 1, interleukin-10 receptor subunit alpha |
| Contenu et stockage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Espèces | Mouse |
| Quantité | 10 μg |
For Reasearch Use Only. Not for use in Humans.